Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Etylenglykol
Etylenglykol ≥99%, clear, colourless liquid
Etylenglykol ≥99%, clear, colourless liquid
:
MP Biomedicals
Etylenglykol ≥99%, clear, colourless liquid
:
MP Biomedicals
:
107-21-1
:
:
HOCH₂CH₂OH
:
MFCD00002885
:
62.07 g/mol
:
3082
:
9,III
:
GHS07