Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Logg inn
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Hjem
Phenol red
Phenol red, powder ACS
Phenol red, powder ACS
:
MP Biomedicals
Phenol red, powder ACS
:
MP Biomedicals
:
143-74-8
:
:
C₁₉H₁₄O₅S
:
MFCD00003552
:
354.38 g/mol
:
205-609-7
:
GHS07
:
Ambient
Identity test :
Passes
pH Transition Intervals::
Yellow 6.8 Orange 7.0 Red 8.2
Clarity of solution:
Passes