Order Entry
Norway
ContactUsLinkComponent
Gallic acid monohydrate, powder ACS
Gallic acid monohydrate, powder ACS
Supplier:  MP Biomedicals
Certificates
Gallic acid monohydrate, powder ACS
Supplier:  MP Biomedicals
Certificates

Specification Test Results

  • Synonyms:
    3,4,5-Trihydroxybenzoic acid
  • Form:
    White Powder
  • Storage:
    Store at Room Temperature (15 to 30 °C), Desiccate
  • Assay:
    ≥98,0%
  • Insoluble Matter:
    ≤0,01%
  • Residue on Ignition:
    ≤0,05%
  • Sulphate (SO4):
    ≤0,02%

Specification Test Results

Related Information