Order Entry
Norway
ContactUsLinkComponent
Ammonium carbonate, white powder
Ammonium carbonate, white powder
Catalog # ICNA0215010880
Supplier:  MP Biomedicals
CAS Number:  
Ammonium carbonate, white powder
Catalog # ICNA0215010880
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Identity Test:
    Passes

Specification Test Results

About this item

 :
 :96.09 g/mol
Melting Pt:58 °C (dec.)
Storage Temperature:Ambient
CAS Number:10361-29-2
 :8000-73-5
Formula:(NH₄)₂CO₃
MDL Number:MFCD00010890