
Ammoniumhydrogenkarbonat, white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:1066-33-7
:
Formula:NH₄HCO₃
MDL Number:MFCD00012138
:79.06 g/mol
:GHS07
Storage Temperature:Romtemperatur
Specification Test Results
- Identity Test:Passes
- pH (1% aq soln) :7.6 - 8.1