kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
Tricaprin
Tricaprin, white solid
Tricaprin, white solid
:
MP Biomedicals
Tricaprin, white solid
:
MP Biomedicals
:
621-71-6
:
:
C₃₃H₆₂O₆
:
MFCD00036239
:
554.85 g/mol
:
Freezer
Identity Test:
Passes