Starch, soluble (from rice), white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:9005-84-9
:
Formula:(C₆H₁₀O₅)n
MDL Number:MFCD00082026
:162.14 g/mol
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- Source:Rice