kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
Tin (II) chloride dihydrate
Tin(II) chloride dihydrate, white crystalline powder
Tin(II) chloride dihydrate, white crystalline powder
:
MP Biomedicals
Tin(II) chloride dihydrate, white crystalline powder
:
MP Biomedicals
:
10025-69-1
:
:
SnCl₂·2H₂O
:
MFCD00149863
:
225.65 g/mol
:
3260
:
8,III
:
GHS05
:
GHS07
:
GHS09