Squalene ≥98%, clear, colourless liquid
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:111-02-4
:
Formula:C₃₀H₅₀
MDL Number:MFCD00008912
:410.73 g/mol
Boiling Pt:285 °C (25 mmHg) (lit.)
Melting Pt:–75 °C (lit.)
Density:0.858 g/mL (25 °C) (lit.)
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- Purity:≥98%
- FTIR:Conforms to Standard