

Collodion 4%, SAFC®
Catalog # 1.02644.1000
Supplier: MERCK PRODUCTION CHEMICALS
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Explosion Limit:1,7 - 36% (V)
- Ignition Temperature:170 °C
- Vapor Pressure:576 hPa (20 °C)
- Storage:Store at 15 to 25 °C
- Appearance:Colorless to Faint Yellowish Clear to Opalescent Viscous Liquid
- Assay:4,0 - 4,2%
- Identity (IR-spectrum):Passes Test
- Neutral Reaction:Passes Test
- Density (d 20 °C/ 4 °C):0,750 - 0,770 g/ml
Specification Test Results
About this item
:
Density:0.76 g/cm³ (20 °C)
Flash Pt:−40 °C
Storage Temperature:Ambient
UN:2059
ADR:3,I
:GHS02
:GHS07
CAS Number:9004-70-0
MDL Number:MFCD00081525