Order Entry
Norway
ContactUsLinkComponent
Puromycin dihydrochloride
Puromycin dihydrochloride
Supplier:  MP Biomedicals
Certificates
Puromycin dihydrochloride
Supplier:  MP Biomedicals

Specification Test Results

  • Synonyms:
    6-dimethylamino-9-(3'-deoxy-3'-(p-methoxy-L-phenylalanyamino)- B-D-ribofuranosyl)-purine
  • Form:
    Off-white Powder
  • Storage:
    Store at -0 °C
  • Extinction Coefficient:
    20,3 (275 nm; 0,1 N NaOH); 19,5 (268 nm, 0,1 NHCl) (Lit.)
  • pka:
    6,8; 7,2 (Lit.)
  • Source:
    Streptomyces albo-niger

Specification Test Results