Adonitol, white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:488-81-3
:
Formula:HOH₂C(CH(OH))₃CH₂OH
MDL Number:MFCD00064291
:152.15 g/mol
Specification Test Results
- Identity Test:Passes
- Melting Point:98 - 106 °C
- FTIR:Conforms to Standard
- Moisture Content:< 0.50%