Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Potassium chloride
Potassium chloride ≥99%, crystalline powder
Potassium chloride ≥99%, crystalline powder
Potassium chloride ≥99%, crystalline powder
Supplier:
MP Biomedicals
Certificates
Expand image
Potassium chloride ≥99%, crystalline powder
Supplier:
MP Biomedicals
About this item
CAS Number:
7447-40-7
:
Formula:
KCl
MDL Number:
MFCD00011360
:
74.55 g/mol
Storage Temperature:
Ambient