Western blotting kit, Perfect Protein™, Novagen®
Catalog # 69078-3
Supplier: Merck Millipore (Novagen)
CAS Number:
Specifications
- Pk:1
- Description:Perfect Protein™ HRP Western Blot Kit, sufficient for 25 blots
Specifications
About this item
The Perfect Protein™ HRP Western Blot Kit includes an HRP or AP conjugate in addition to Perfect Protein™ Western markers.
- S-protein conjugate (included in kits) does not interfere with antibodies or streptavidin detection
- Can be used with colorimetric and chemiluminescent AP or HRP substrates
- Recombinant, unmodified markers give sharp, accurately sized bands
- Markers are supplied at the working dilution in gel loading buffer; concentration optimised for Western detection
- Protein sizes are 15, 25, 35, 50, 75, 100, and 150 kDa