Anti-EPCAM/1110 Mouse Monoclonal Antibody [clone: EGP40/1110]
Catalog # ABNOMAB13327
Supplier: Abnova
Specifications
- Antistofftype:Primary
- Antigensymbol:EPCAM/1110
- Klonalitet:Monoclonal
- Klon:EGP40/1110
- Vert:Mouse
- ImmunoKjemi:Yes
- Isotype:IgG2b kappa
- Reaktivitet:Human,Rat
- Gen-ID:4072
- Str:100 μg
- Fp:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant human EPCAM.
Type: Primary
Antigen: EPCAM/1110
Clonality: Monoclonal
Clone: EGP40/1110
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human,Rat