Order Entry
Norway
ContactUsLinkComponent
Anti-CD24 Rat Monoclonal Antibody (PE) [clone: M1/69]
Anti-CD24 Rat Monoclonal Antibody (PE) [clone: M1/69]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-CD24 Rat Monoclonal Antibody (PE) [clone: M1/69]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 50-0242-U100
  • 50-0242-U025
  • Antigen symbol:
  • CD24
  • CD24
  • Antigen name:
  • CD24, Heat Stable Antigen
  • CD24, Heat Stable Antigen
  • Antigen synonyms:
  • Heat Stable Antigen|HSA
  • Heat Stable Antigen|HSA
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • M1/69
  • M1/69
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 12484
  • 12484
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • +2 °C to +8 °C, protected from light
  • +2 °C to +8 °C, protected from light

Specifications

Related Information