Test kits, photometric, AccuVac®
Leverandør: Hach
Sertifikat
Om dette produktet
AV - AccuVac® - Sealed glass cuvettes containing a portioned amount of reagent. Simply immerse the tip of the accuvac in the sample, break it and the vacuum draws the sample into the cuvette.