Order Entry
Norway
ContactUsLinkComponent
Standards for colorimeters
Standards for colorimeters
Catalog # HACHLZM268
Supplier:  Hach
CAS Number:  
undefined
Standards for colorimeters
Catalog # HACHLZM268
Supplier:  Hach
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    Black/white standard
Specifications
About this item
These standards are used for the calibration of colorimeters.
Certifications: Certified in accordance with DIN 5033.