pH meter, handheld, SD 50
Catalog # LOVI194800
Supplier: Lovibond
Specifications
- Temperature range:0...+60 °C
- Description:pH meter, handheld, SD 50
- Seq:01
- Model:SD 50
- Temperature accuracy:±1 °C
- Temperature resolution:0,1 °C
- Compliance:IP 67
- Data logging capacity:25 data sets (date/time/temperature/result)
- Display:LCD (22×22 mm), with backlight
- Power supply:2 × CR2032 batteries (>25 hours continuous use)
- Temperature compensation:Automatic
- Weight:0,10 kg
- pH accuracy:±0,05
- pH calibration:1-, 2- or 3-point (with auto-recognition (NIST / IUPAC))
- pH range:0 - 14
- pH resolution:0,01
- Pk:1
Specifications
About this item
The SD 50 pH meter is a compact, easy to use handheld meter for the measurement of pH.
- Intuitive scroll-through functionality
- Robust plastic case with hanger
- Data storage function for 25 sets of data, each with date and time stamp
- Backlit display
- Selectable buffer system: pH 7,00 or pH 6,86
With robust housing and fully waterproof (IP67) casing, this tester is the ideal solution for in-situ testing in environmental, industrial or pool & spa applications. The series can be upgraded with replaceable electrodes to ensure long-life functionality in the field.
Delivery: Meter is supplied with two batteries, lanyard, robust carrying case and instruction manual.