Order Entry
Norway
ContactUsLinkComponent
Test kits, AQUALYTIC®
Test kits, AQUALYTIC®
Catalog # LOVI418453
Supplier:  Lovibond
CAS Number:  
undefined
Test kits, AQUALYTIC®
Catalog # LOVI418453
Supplier:  Lovibond
CAS Number:  

Specifications

  • Pk:
    1
  • Description:
    Boiler water-set KW-3, for alkalinity (p- + m-value), residual hardness, phosphate (ortho), pH-value and sulphite
  • Seq:
    02
  • Type:
    Titrimetry method(Alkalinity, residual hardness and sulphite); Colorimetry method(Phosphate (ortho)); Indicator strips(pH)
  • Range:
    Alkalinity: 1 drop = 1 or 0,5 mmol/l;
    Residual hardness: 1 drop = 0,1 or 0,05 °dH;
    Ortho phosphate: 2,5 - 25 mg/l PO43–;
    pH: 7,0 - 14;
    Sulphite: 1 drop = 5 or 2,5 mg/l Na2SO3

Specifications

About this item

These kits are mainly designed for water testing (boiling, cooling and domestic water).

  • Rapid quantitative determination
  • For field and laboratory tests
  • Economically priced through refill
  • Both colorimetric and titrimetric methods are used

The detailed instructions contain a step-by-step explanation of the test procedure.

Delivery: Kits are supplied in a sturdy, compact plastic case containing all the necessary chemicals and reagents in liquid or powder form to conduct the tests.

Refill packs

Product Information