Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromoimidazole
2-Bromoimidazole 98%
2-Bromoimidazole 98%
:
Thermo Fisher Scientific
2-Bromoimidazole 98%
:
Thermo Fisher Scientific
:
16681-56-4
:
:
C₃H₃BrN₂
:
MFCD02179526
:
146.97 g/mol
:
189…190 °C
:
GHS05
:
GHS07
:
Refrigerator
Appearance:
White to light brown Powder
Infrared spectrum:
Conforms
HPLC:
≥97.5 %
NMR (1H-NMR):
Consistent with structure
ACRO45463