Order Entry
Norway
ContactUsLinkComponent
Pyridine 99.8%, AcroSeal® for biochemistry
Pyridine 99.8%, AcroSeal® for biochemistry
Catalog # 447900010.
CAS Number:  
undefined
Pyridine 99.8%, AcroSeal® for biochemistry
Catalog # 447900010.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.05 %
  • GC:
    ≥99.75 %
  • UV:
    (neat)
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.0005 %
  • Chloride (Cl):
    ≤0.0005 %
  • Sulfate (SO4):
    ≤0.0005 %
  • Cadmium (Cd):
    ≤0.000005 %
  • Copper (Cu):
    ≤0.000002 %
  • Ammonia (NH3):
    ≤0.002 %

Specification Test Results

About this item

 :
Boiling Pt:115…116 °C (760 mmHg)
Melting Pt:–42 °C
Density:0.978 g/cm³
Flash Pt:17 °C
Storage Temperature:Ambient
UN:1282
ADR:3,II
 :79.1 g/mol
 :GHS07
 :GHS02
CAS Number:110-86-1
Formula:C₅H₅N
MDL Number:MFCD00011732