Gallic acid monohydrate ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5995-86-8
:
Formula:C₇H₆O₅·1H₂O
MDL Number:MFCD00149098
:188.14 g/mol
Melting Pt:251 °C (dec.)
Flash Pt:250 °C (closed cup)
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to beige Crystalline powder
- Infrared spectrum:Conforms
- Residue after ignition:≤0.05 %
- Titration with NaOH:≥98.0 %
- Sulfate (SO4):≤0.02
- Insoluble matter:≤0.01 %