
Ammonium hydrogen sulphate, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7803-63-6
:
Formula:H₅NO₄S
MDL Number:MFCD00051073
:115.11 g/mol
Boiling Pt:350 °C (760 mmHg)
Melting Pt:147 °C (dec.)
Density:1,787 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:2506
ADR:8,II
:GHS05
Specification Test Results
- Appearance:White Crystalline powder
- Titration with NaOH:≥97.5 %
- Chloride (Cl):≤0.005 %
- Phosphate (PO4):≤0.003 %
- Insoluble matter:≤0.05 %