

Ethanol 96%, Technical Grade (not denatured)
Catalog # ACRO444190050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Plastic drum
- Pk:5 L
- Seq:5190
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Refractive index:1.3610 to 1.3640 (20°C, 589 nm)
- Density:(20°C) 0.803 to 0.812 kg/l
- Ethanol:95 to 97 % v/v (calculated)
Specification Test Results
About this item
:
:46.07 g/mol
Melting Pt:–114 °C
Density:0.8 g/cm³
Flash Pt:17 °C
UN:1170
ADR:3,II
:GHS02
:GHS07
Storage Temperature:Ambient
CAS Number:64-17-5
Formula:C₂H₅OH
MDL Number:MFCD00003568