1,2-Dimethoxyethane, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:110-71-4
:
Formula:C₄H₁₀O₂
MDL Number:MFCD00008502
:90.12 g/mol
Boiling Pt:85 °C (1013 hPa)
Melting Pt:–69 °C
Density:0.867 g/cm³
Flash Pt:–6 °C
UN:2252
ADR:3,II
:GHS07
:GHS08
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Water:≤0.005 %
- GC:≥99.4 %
- Stabilizer:Approx. 100 ppm BHT