
Boc-N-methyl-D-alanine 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16948-16-6
:
Formula:C₉H₁₇NO₄
MDL Number:MFCD00037242
:203.2 g/mol
Melting Pt:88…92 °C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Specific optical rotation:-29° to -31° (20°C, 589 nm) (c=1 in EtOH)
- Titration with NaOH:98.5 to 101.5 %