Boc-D-aspartic acid β-benzyl ester 98%
Katalog # ACRO442870050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- Specific optical rotation:+18° to +21° (20°C, 589 nm) (c=1 in DMF)
Specification Test Results
About this item
:
:323.345 g/mol
Melting Pt:101…105 °C
Storage Temperature:Refrigerator
CAS Number:51186-58-4
Formula:C₁₆H₂₁NO₆
MDL Number:MFCD00038255