
Di-tert-butyl hydrazodicarboxylate 97%
ACRO440420100
:
- Pack type:Glass bottle
- Pk:10 g
- Seq:3236
- Appearance:White Powder
- Infrared spectrum:Conforms
- GC:≥96.0 %
:
:232.28 g/mol
:120…125 °C
:GHS07
:Ambient
:16466-61-8
:C₁₀H₂₀N₂O₄
:MFCD00015000