Propidium iodide 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:25535-16-4
:
Formula:C₂₇H₃₄I₂N₄
MDL Number:MFCD00011921
:668.4 g/mol
Melting Pt:220…225 °C (dec.)
:GHS07
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Red Powder
- Infrared spectrum:Conforms
- Melting point:200 to 225 °C
- Titration Argentometric:≥94 %
- Water:≤5 %