Tris acetate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6850-28-8
:
Formula:NH₂C(CH₂OH)₃·CH₃COOH
MDL Number:MFCD00038951
:181.1870 g/mol
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder
- Water:≤1.0 %
- Solubility:(1 g/ml in water) Clear colorless
- UV:(50 % in water)
- Titration with NaOH:≥98.5 %
- Heavy metals (as Pb):≤5 ppm