
4-Bromomethyl-2,6-dichloropyridine 97%
Catalog # ACRO433440010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to yellow to brown Crystalline powder or low melting solid
- Infrared spectrum:Conforms
- Melting point:40 to 45 °C
- GC:≥96.0 %
Specification Test Results
About this item
:
:240.909 g/mol
Boiling Pt:118 °C (.2 mmHg)
Melting Pt:42.5…43.2 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
CAS Number:175204-45-2
Formula:C₆H₄BrCl₂N
MDL Number:MFCD00052637