2-Amino-5-thiazolecarboxylic acid 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:40283-46-3
:
Formula:C₄H₄N₂O₂S
MDL Number:MFCD06203554
:144.1500 g/mol
Melting Pt:182…184 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Off-white to yellow Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥94 %
- Color scale:≤10 APHA
- Non-volatile matter:≤0.001 %