Order Entry
Norway
ContactUsLinkComponent
Boron trichloride 10% (w/w) 1 M in 2-chloroethanol, AcroSeal®
Toxic
Corrosive
Boron trichloride 10% (w/w) 1 M in 2-chloroethanol, AcroSeal®
Catalog # 428331000.
CAS Number:  
undefined
Toxic
Corrosive
Boron trichloride 10% (w/w) 1 M in 2-chloroethanol, AcroSeal®
Catalog # 428331000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Titration Argentometric:
    0.9 to 1.1 M
  • Melting point:
    62 to 66 °C
  • GC:
    ≥96.0 %

Specification Test Results

About this item

 :
 :117.17 g/mol
Density:1.26 g/cm³
Flash Pt:53 °C
Storage Temperature:Refrigerator
UN:3390
ADR:6.1,
 :GHS06
 :GHS05
CAS Number:10294-34-5
Formula:BCl₃
MDL Number:MFCD00011313