Order Entry
Norway
ContactUsLinkComponent
 
Irritant
4-Hydroxy-3-methylbenzaldehyde 98%
Catalog # ACRO427550010
CAS Number:  
undefined
Irritant
4-Hydroxy-3-methylbenzaldehyde 98%
Catalog # ACRO427550010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Off-white to tan to brown Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    118 to 120 °C
  • GC:
    ≥97.5 %
  • NMR (1H-NMR):
    Consistent with structure

Specification Test Results

About this item

 :
Melting Pt:118…120 °C
 :136.1479 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:15174-69-3
Formula:C₈H₈O₂
MDL Number:MFCD00012360