kr 0,00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Norway
ContactUsLinkComponent
3-Chloro-4-formylpyridine
3-Chloro-4-formylpyridine 97%
3-Chloro-4-formylpyridine 97%
:
Thermo Fisher Scientific
3-Chloro-4-formylpyridine 97%
:
Thermo Fisher Scientific
:
72990-37-5
:
:
C₆H₄ClNO
:
MFCD06200712
:
141.56 g/mol
:
225 °C
:
67…68 °C
:
GHS07
:
Refrigerator
Appearance:
White to yellow Crystalline powder
Infrared spectrum:
Conforms
GC:
≥96.0 %
ACRO42655