Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
kr 0,00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Norway
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(Ethoxycarbonyl)phenylboronic acid
2-(Ethoxycarbonyl)phenylboronic acid 97%
2-(Ethoxycarbonyl)phenylboronic acid 97%
:
Thermo Fisher Scientific
2-(Ethoxycarbonyl)phenylboronic acid 97%
:
Thermo Fisher Scientific
:
380430-53-5
:
:
C₉H₁₁BO₄
:
MFCD02179453
:
193.99 g/mol
:
137…138 °C
:
GHS07
:
Ambient
Appearance:
White to off-white Solid
Infrared spectrum:
Conforms
HPLC:
≥96.0 %
NMR (1H-NMR):
Consistent with structure
ACRO39383