Sodium chloride ∼26% in water, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7647-14-5
:
Formula:NaCl
MDL Number:MFCD00003477
:58.44 g/mol
Density:1.202 g/cm³
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless Solution
- Titration Argentometric:25 to 28 %