
4-(Trifluoromethyl)hydrocinnamic acid 95%
:53473-36-2
:
:C₁₀H₉F₃O₂
:MFCD00674032
:218.17 g/mol
:106…110 °C
:GHS07
:Ambient
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:111 to 119 °C
- HPLC:≥94 %