

2-Cyanoethyl-N,N-diisopropylchlorophosphoramidite 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:89992-70-1
:
Formula:C₉H₁₈ClN₂OP
MDL Number:MFCD00011544
:236.68 g/mol
Density:1.061 g/cm³
Flash Pt:110 °C
Storage Temperature:Freezer
UN:2845
ADR:4.2,I
:GHS02
:GHS05
Specification Test Results
- Appearance:Clear to hazy colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4760 to 1.4800 (20°C, 589 nm)
- Chloride (Cl):14.5 to 15.5 %