tert-Butyl (3S)-3-amino-3-phenylpropanoate 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:120686-18-2
:
Formula:C₁₃H₁₉NO₂
MDL Number:MFCD00798310
:221.3 g/mol
Storage Temperature:Freezer
UN:2735
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Clear colorless to light brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4960 to 1.4990 (20°C, 589 nm)
- Specific optical rotation:-17° to -22° (20°C, 589 nm) (c=1, CHCl3)
- Titration with HClO4:≥96.0 %