Order Entry
Norway
ContactUsLinkComponent
 
 
 
1-Chloro-N,N-2-trimethylpropenylamine 98.5+%
 
undefined
 
 
1-Chloro-N,N-2-trimethylpropenylamine 98.5+%

 

  • Appearance:
    Colorless to yellow Liquid (May develop some turbidity or precipitate)
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4520 to 1.4560 (20°C, 589 nm)
  • Titration Argentometric:
    98.50 to 101.50 % (ex Cl)