Boc-N-2-dimethylalanine 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:146000-39-7
:
Formula:C₁₀H₁₉NO₄
MDL Number:MFCD01862912
:217.265 g/mol
Melting Pt:153…157 °C
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:153 to 157 °C
- Titration with NaOH:≥98.0 %