
Sodium glycocholate hydrate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:207614-05-9
:338950-81-5
Formula:C₂₆H₄₂NO₆Na·xH₂O
MDL Number:MFCD09037360
:487.60 g/mol (anhy)
Melting Pt:210…215 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to cream Powder
- Infrared spectrum:Conforms
- Water:≤8 % (K.F.)
- HPLC:≥97.5 %
- Specific optical rotation:+28° to +31° (20°C, 589 nm) (c=1, H2O)
- Titration with HClO4:97.5 to 102.5 % (On anhydrous basis)