Dowex® 1X2 (Cl) 200-400 mesh ion-exchange resin
Catalog # ACRO202971000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Appearance:Beads
- Particle size:200 to 400 mesh
- Wet volume capacity:≥0.6 meq/ml (Cl)
- Water retention capacity:65 to 75 % (Cl)
Specification Test Results
About this item
:
Storage Temperature:Ambient
CAS Number:9085-42-1
:69011-19-4
MDL Number:MFCD00132715