


Cinnamyl chloride 95% (trans-isomers)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2687-12-9
:
Formula:C₉H₉Cl
MDL Number:MFCD00000986
:152.62 g/mol
Boiling Pt:120…126 °C (15 mmHg)
Melting Pt:7 °C
Density:1.096 g/cm³
Flash Pt:79 °C
UN:2922
ADR:8,II
:GHS05
:GHS06
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to yellow to yellow-brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5830 to 1.5850 (20°C, 589 nm)
- GC:≥94 %
- HPLC:≥97.0 %