Order Entry
Norway
ContactUsLinkComponent
1,3-Bis(aminomethyl)cyclohexane (cis and trans mixture) 99%
Irritant
Corrosive
1,3-Bis(aminomethyl)cyclohexane (cis and trans mixture) 99%
Certificates
undefined
Irritant
Corrosive
1,3-Bis(aminomethyl)cyclohexane (cis and trans mixture) 99%

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4920 to 1.4940 (20°C, 589 nm)
  • GC:
    ≥98.5 %

Specification Test Results

Related Information