Aleuritic acid 95%
Catalog # ACRO133730250
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to beige Crystalline powder
- Infrared spectrum:Conforms
- Titration with NaOH:≥94 %
- GC:≥96.0 %
Specification Test Results
About this item
:
:304.43 g/mol
Melting Pt:100…105 °C
Storage Temperature:Ambient
CAS Number:533-87-9
Formula:C₁₆H₃₂O₅
MDL Number:MFCD00135596