Methyl (2E)-pent-2-enoate 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15790-88-2
:
Formula:C₆H₁₀O₂
MDL Number:MFCD00137611
:114.144 g/mol
Flash Pt:20 °C
UN:1993
ADR:3,II
:GHS07
:GHS02
Storage Temperature:Freezer
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4300 to 1.4320 (20°C, 589 nm)
- GC:≥94 %