Order Entry
Norway
ContactUsLinkComponent
1-Adamantyl isocyanate 98%
1-Adamantyl isocyanate 98%
Certificates
undefined
1-Adamantyl isocyanate 98%

Specification Test Results

  • Appearance:
    White Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    140 to 150 °C
  • GC:
    ≥97.5 %

Specification Test Results

Related Information